6-Chloro-1,2,3,4-tetrahydropyrido[2,3-b]pyrazine ≥95%

Supplier: Matrix Scientific

073757-250MG 073757-1G
102531-838EA 446.15 USD
102531-838 102531-840
6-Chloro-1,2,3,4-tetrahydropyrido[2,3-b]pyrazine ≥95%
6-Chloro-1,2,3,4-tetrahydropyrido[2,3-b]pyrazine

Hazard: Irritant

Formula: C₇H₈ClN₃
MDL Number: MFCD08062760
CAS Number: 1210129-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR