3-Chloroanthranilic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-3-chlorobenzoic acid

073754-100G 073754-25G
102531-828EA 180 USD
102531-828 102833-652
3-Chloroanthranilic acid ≥95%
3-Chloroanthranilic acid

Hazard: Irritant/Light Sensitive/Air Sensitive

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 189-193°C
MDL Number: MFCD00134229
CAS Number: 6388-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR