5-Methoxypyrimidine ≥95%

Supplier: Matrix Scientific

073753-250MG 073753-1G 073753-5G
102531-818EA 355.38 USD
102531-818 102531-820 102531-822
5-Methoxypyrimidine ≥95%
5-Methoxypyrimidine

Hazard: Irritant

Formula: C5H6N2O
MDL Number: MFCD04971313
CAS Number: 31458-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR