5-((Trimethylsilyl)ethynyl)pyrimidine ≥95%
Supplier: Matrix Scientific
073752-250MG
073752-1G
073752-5G
102531-812EA
498.46
USD
102531-812
102531-814
102531-816
5-((Trimethylsilyl)ethynyl)pyrimidine ≥95%
5-((Trimethylsilyl)ethynyl)pyrimidine
Hazard: Irritant
Formula:
C₉H₁₂N₂Si MW: 176.2900 g/mol |
MDL Number:
MFCD09258881 CAS Number: 216309-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...