7-Bromo-1,2-dihydroisoquinolin-3(4H)-one ≥95%

Supplier: Matrix Scientific

073750-1G 073750-5G
102531-802EA 515.38 USD
102531-802 102531-804
7-Bromo-1,2-dihydroisoquinolin-3(4H)-one ≥95%
7-Bromo-1,2-dihydroisoquinolin-3(4H)-one

Hazard: Irritant

Formula: C₉H₈BrNO
MDL Number: MFCD13193419
CAS Number: 943751-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR