2-(4-Methylpiperazin-1-yl)pyrimidin-5-amine ≥95%

Supplier: Matrix Scientific

073747-250MG 073747-1G
102531-788EA 541.54 USD
102531-788 102531-790
2-(4-Methylpiperazin-1-yl)pyrimidin-5-amine ≥95%
2-(4-Methylpiperazin-1-yl)pyrimidin-5-amine

Hazard: Irritant

Formula: C₉H₁₅N₅
MDL Number: MFCD13193411
CAS Number: 943757-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR