2-Chloro-4-ethynylpyridine ≥95%

Supplier: Matrix Scientific

073746-250MG 073746-1G 073746-5G
102531-782EA 498.46 USD
102531-782 102531-784 102531-786
2-Chloro-4-ethynylpyridine ≥95%
2-Chloro-4-ethynylpyridine

Hazard: Irritant

Formula: C₇H₄ClN
MW: 137.5664 g/mol
MDL Number: MFCD13189742
CAS Number: 945717-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR