7-Bromo-2-chlorothieno[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

073745-250MG 073745-1G
102531-778EA 541.54 USD
102531-778 102531-780
7-Bromo-2-chlorothieno[3,2-d]pyrimidine ≥95%
7-Bromo-2-chlorothieno[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₆H₂BrClN₂S
MW: 249.5200 g/mol
MDL Number: MFCD13193398
CAS Number: 1152475-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR