4-Bromo-2-nitrobenzonitrile ≥95%
Supplier: Matrix Scientific
073744-1G
073744-5G
073744-10G
102531-772EA
427.69
USD
102531-772
102531-774
102531-776
4-Bromo-2-nitrobenzonitrile ≥95%
4-Bromo-2-nitrobenzonitrile
Hazard: Irritant
Formula:
C₇H₃BrN₂O₂ |
MDL Number:
MFCD00087225 CAS Number: 79603-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...