4'-Cyano-3'-fluorobiphenyl-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

073743-250MG 073743-1G
102531-768EA 740 USD
102531-768 102531-770
4'-Cyano-3'-fluorobiphenyl-4-carboxylic acid ≥95%
4'-Cyano-3'-fluorobiphenyl-4-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₈FNO₂
CAS Number: 1260811-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR