5-Aminonicotinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-3-pyridinecarbonitrile

073740-1G 073740-5G
102531-754EA 307.69 USD
102531-754 102531-756
5-Aminonicotinonitrile ≥95%
5-Aminonicotinonitrile

Hazard: Irritant

Formula: C₆H₅N₃
MW: 119.1200 g/mol
MDL Number: MFCD07367907
CAS Number: 13600-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR