6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one ≥95%

Supplier: Matrix Scientific

073739-250MG 073739-1G
102531-750EA 767.69 USD
102531-750 102531-752
6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one ≥95%
6-Chloropyrido[3,4-d]pyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₇H₄ClN₃O
MW: 181.5800 g/mol
MDL Number: MFCD11044635
CAS Number: 171178-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR