6-Bromo-3-chloropyrazin-2-amine ≥95%

Supplier: Matrix Scientific

073737-250MG 073737-1G 073737-5G
102531-740EA 450.42 USD
102531-740 102531-742 102531-744
6-Bromo-3-chloropyrazin-2-amine ≥95%
6-Bromo-3-chloropyrazin-2-amine

Hazard: Irritant

Formula: C₄H₃BrClN₃
MW: 208.4400 g/mol
MDL Number: MFCD13193368
CAS Number: 1082843-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR