Methyl-2-amino-5-bromopyrimidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-5-bromopyrimidine-4-carboxylate

073734-250MG 073734-1G 073734-5G
102531-724EA 283.08 USD
102531-724 102531-726 102531-728
Methyl-2-amino-5-bromopyrimidine-4-carboxylate ≥95%
Methyl-2-amino-5-bromopyrimidine-4-carboxylate

Hazard: Irritant

Formula: C₆H₆BrN₃O₂
MW: 232.0400 g/mol
MDL Number: MFCD09966084
CAS Number: 1034737-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR