5-Iodo-4-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073733-250MG 073733-1G
102531-720EA 1821.54 USD
102531-720 102531-722
5-Iodo-4-methylpyrimidine ≥95%
5-Iodo-4-methylpyrimidine

Hazard: Irritant

Formula: C5H5IN2
CAS Number: 91749-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR