1-(6'-Aminospiro[cyclopropane-1,3'-indolin]-1'-yl)ethanone ≥95%

Supplier: Matrix Scientific

073732-250MG 073732-1G
102531-716EA 826.15 USD
102531-716 102531-718
1-(6'-Aminospiro[cyclopropane-1,3'-indolin]-1'-yl)ethanone ≥95%
1-(6'-Aminospiro[cyclopropane-1,3'-indolin]-1'-yl)ethanone

Hazard: Irritant

Formula: C₁₂H₁₄N₂O
MW: 202.250 g/mol
MDL Number: MFCD13195332
CAS Number: 645419-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR