5'-Fluorospiro[cyclopropane-1,3'-indoline] ≥95%

Supplier: Matrix Scientific

073731-250MG 073731-1G
102531-712EA 681.04 USD
102531-712 102531-714
5'-Fluorospiro[cyclopropane-1,3'-indoline] ≥95%
5'-Fluorospiro[cyclopropane-1,3'-indoline]

Hazard: Irritant

Formula: C10H10FN
CAS Number: 913179-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR