5-Bromobenzo[d]isothiazol-3-amine ≥95%

Supplier: Matrix Scientific

073729-250MG 073729-1G
102531-702EA 1841.54 USD
102531-702 102531-704
5-Bromobenzo[d]isothiazol-3-amine ≥95%
5-Bromobenzo[d]isothiazol-3-amine

Hazard: Irritant

Formula: C₇H₅BrN₂S
CAS Number: 613262-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR