Methyl-4-aminopyrimidine-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-aminopyrimidine-5-carboxylate

073728-250MG 073728-1G 073728-5G
102531-696EA 320 USD
102531-696 102531-698 102531-700
Methyl-4-aminopyrimidine-5-carboxylate ≥95%
Methyl-4-aminopyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₆H₇N₃O₂
MW: 153.1400 g/mol
MDL Number: MFCD13195321
CAS Number: 714975-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR