2-Methylquinolin-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-2-methylquinoline dihydrochloride

073727-250MG 073727-1G
102531-692EA 421.18 USD
102531-692 102531-694
2-Methylquinolin-3-amine ≥95%
2-Methylquinolin-3-amine

Hazard: Irritant

Formula: C₁₀H₁₀N₂
MW: 158.1998 g/mol
MDL Number: MFCD08705651
CAS Number: 21352-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR