7-Bromothieno[2,3-b]pyrazine ≥95%

Supplier: Matrix Scientific

073724-250MG 073724-1G
102531-678EA 1395.38 USD
102531-678 102531-680
7-Bromothieno[2,3-b]pyrazine ≥95%
7-Bromothieno[2,3-b]pyrazine

Hazard: Irritant

Formula: C₆H₃BrN₂S
MW: 215.07 g/mol
MDL Number: MFCD13193345
CAS Number: 1126824-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR