Thieno[2,3-b]pyrazine ≥95%

Supplier: Matrix Scientific

073723-250MG 073723-1G
102531-674EA 826.15 USD
102531-674 102531-676
Thieno[2,3-b]pyrazine ≥95%
Thieno[2,3-b]pyrazine

Hazard: Irritant

Formula: C₆H₄N₂S
CAS Number: 56088-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR