3-Bromopyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

073719-250MG 073719-1G 073719-5G
102531-656EA 229.23 USD
102531-656 102531-658 102531-660
3-Bromopyrazine-2-carboxylic acid ≥95%
3-Bromopyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C5H3BrN2O2
MW: 202.9900 g/mol
MDL Number: MFCD08753934
CAS Number: 937669-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR