Methyl-3-bromopyrazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-Bromo-2-pyrazinecarboxylate

073718-250MG 073718-1G 073718-5G
102531-650EA 2061.54 USD
102531-650 102531-652 102531-654
Methyl-3-bromopyrazine-2-carboxylate ≥95%
Methyl-3-bromopyrazine-2-carboxylate

Hazard: Irritant

Formula: C₆H₅BrN₂O₂
MW: 217.0200 g/mol
MDL Number: MFCD08753935
CAS Number: 51171-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR