(4-Bromopyridin-2-yl)methanamine ≥95%

Supplier: Matrix Scientific

073716-250MG 073716-1G
102531-642EA 769.23 USD
102531-642 102531-644
(4-Bromopyridin-2-yl)methanamine ≥95%
(4-Bromopyridin-2-yl)methanamine

Hazard: Irritant

Formula: C₆H₇BrN₂
MDL Number: MFCD07374903
CAS Number: 865156-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR