tert-Butyl (4-bromothiophen-2-yl)carbamate ≥95%

Supplier: Matrix Scientific

073713-250MG 073713-1G 073713-5G
102531-624EA 541.54 USD
102531-624 102531-626 102531-628
tert-Butyl (4-bromothiophen-2-yl)carbamate ≥95%
tert-Butyl (4-bromothiophen-2-yl)carbamate

Hazard: Irritant

Formula: C₉H₁₂BrNO₂S
MDL Number: MFCD13195314
CAS Number: 868387-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR