7-Chloro[1,2,4]triazolo[1,5-c]pyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Chloro-[1,2,4]triazolo[1,5-c]pyrimidine

073712-250MG 073712-1G
102531-620EA 1066.15 USD
102531-620 102531-622
7-Chloro[1,2,4]triazolo[1,5-c]pyrimidine ≥95%
7-Chloro[1,2,4]triazolo[1,5-c]pyrimidine

Hazard: Irritant

Formula: C₅H₃ClN₄
MW: 154.5571 g/mol
MDL Number: MFCD11043978
CAS Number: 1159811-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR