2-Chloro-7-fluoro-1H-benzo[d]imidazole ≥95%
Supplier: Matrix Scientific
073711-250MG
073711-1G
073711-5G
102531-614EA
640
USD
102531-614
102531-616
102531-618
2-Chloro-7-fluoro-1H-benzo[d]imidazole ≥95%
2-Chloro-7-fluoro-1H-benzo[d]imidazole
Hazard: Irritant
Formula:
C₇H₄ClFN₂ |
CAS Number:
256519-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...