3-Bromo-5-fluoropyridin-4-amine ≥95%
Supplier: Matrix Scientific
073707-250MG
073707-1G
073707-5G
102531-594EA
569.23
USD
102531-594
102531-596
102531-598
3-Bromo-5-fluoropyridin-4-amine ≥95%
3-Bromo-5-fluoropyridin-4-amine
Hazard: Irritant
Formula:
C5H4BrFN2 |
MDL Number:
MFCD13185327 CAS Number: 1214326-89-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...