5-Bromo-3-fluoropyridin-2-amine ≥95%

Supplier: Matrix Scientific

073705-1G 073705-5G
102531-584EA 130.77 USD
102531-584 102531-586
5-Bromo-3-fluoropyridin-2-amine ≥95%
5-Bromo-3-fluoropyridin-2-amine

Hazard: Irritant/Refrigerate

Formula: C₅H₄BrFN₂
Melting Pt: 54…56 °C
MDL Number: MFCD11848483
CAS Number: 748812-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR