2-Amino-5-chlorobenzaldehyde ≥95%

Supplier: Matrix Scientific

073704-1G 073704-5G
102531-578EA 123.08 USD
102531-578 102531-580
2-Amino-5-chlorobenzaldehyde ≥95%
2-Amino-5-chlorobenzaldehyde

Hazard: Irritant/Air Sensitive/Refrigerate

Formula: C₇H₆ClNO
MW: 155.58 g/mol
Melting Pt: 68-74°C
MDL Number: MFCD02093936
CAS Number: 20028-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR