Ethyl-2-piperazin-1-ylthiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

073703-250MG 073703-1G 073703-5G
102531-572EA 569.23 USD
102531-572 102531-574 102531-576
Ethyl-2-piperazin-1-ylthiazole-4-carboxylate ≥95%
Ethyl-2-piperazin-1-ylthiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₂S
CAS Number: 104481-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR