Ethyl-2-(4-(tert-butoxycarbonyl)piperazin-1-yl)thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

073702-250MG 073702-1G 073702-5G
102531-566EA 400 USD
102531-566 102531-568 102531-570
Ethyl-2-(4-(tert-butoxycarbonyl)piperazin-1-yl)thiazole-4-carboxylate ≥95%
Ethyl-2-(4-(tert-butoxycarbonyl)piperazin-1-yl)thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₃N₃O₄S
MDL Number: MFCD08445626
CAS Number: 867065-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR