3-Bromo-7-fluoro-1H-indazole ≥95%

Supplier: Matrix Scientific

073701-250MG 073701-1G 073701-5G
102531-560EA 640 USD
102531-560 102531-562 102531-564
3-Bromo-7-fluoro-1H-indazole ≥95%
3-Bromo-7-fluoro-1H-indazole

Hazard: Irritant

Formula: C₇H₄BrFN₂
MW: 215.0200 g/mol
MDL Number: MFCD13193285
CAS Number: 1257853-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR