4,5-Dichloro-6-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073700-1G 073700-5G
102531-556EA 284.62 USD
102531-556 102531-558
4,5-Dichloro-6-methylpyrimidine ≥95%
4,5-Dichloro-6-methylpyrimidine

Hazard: Irritant

Formula: C₅H₄Cl₂N₂
MDL Number: MFCD09909763
CAS Number: 83942-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR