5-(2-Methoxyethoxy)pyrazin-2-amine ≥95%

Supplier: Matrix Scientific

073699-250MG 073699-1G
102531-550EA 1647.69 USD
102531-550 102531-552
5-(2-Methoxyethoxy)pyrazin-2-amine ≥95%
5-(2-Methoxyethoxy)pyrazin-2-amine

Hazard: Irritant

Formula: C₇H₁₁N₃O₂
MW: 169.1800 g/mol
MDL Number: MFCD13193283
CAS Number: 710322-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR