Ethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 7-azaindole-2-carboxylate, Ethyl 1H-pyrrolo[2,3-b]pyridine-2-carboxylate
073697-1G
073697-5G
102531-542EA
361.54
USD
102531-542
102531-544
Ethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥95%
Ethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₂ MW: 190.2000 g/mol |
MDL Number:
MFCD09998715 CAS Number: 221675-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...