5-(Benzyloxy)pyridin-2-amine ≥95%

Supplier: Matrix Scientific

073696-250MG 073696-1G 073696-5G
102531-534EA 456.92 USD
102531-534 102531-536 102531-538
5-(Benzyloxy)pyridin-2-amine ≥95%
5-(Benzyloxy)pyridin-2-amine

Hazard: Irritant

Formula: C₁₂H₁₂N₂O
MW: 200.2400 g/mol
MDL Number: MFCD11135746
CAS Number: 96166-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR