2-Bromo-6-chloro-3-nitropyridine ≥95%

Supplier: Matrix Scientific

073694-1G 073694-5G
102531-524EA 289.23 USD
102531-524 102531-526
2-Bromo-6-chloro-3-nitropyridine ≥95%
2-Bromo-6-chloro-3-nitropyridine

Hazard: Irritant

Formula: C₅H₂BrClN₂O₂
MW: 237.4400 g/mol
MDL Number: MFCD09260904
CAS Number: 91678-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR