exo-8-Benzyl-8-azabicyclo[3.2.1]octan-3-amine ≥95%
Supplier: Matrix Scientific
073693-250MG
073693-1G
073693-5G
102531-518EA
498.46
USD
102531-518
102531-520
102531-522
exo-8-Benzyl-8-azabicyclo[3.2.1]octan-3-amine ≥95%
exo-8-Benzyl-8-azabicyclo[3.2.1]octan-3-amine
Hazard: Irritant
Formula:
C₁₄H₂₀N₂ MW: 216.322 g/mol |
MDL Number:
MFCD09753144 CAS Number: 76272-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...