2-Fluoro-4-iodonicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-4-iodo-nicotinic acid

073692-1G 073692-5G
102531-514EA 121.54 USD
102531-514 102531-516
2-Fluoro-4-iodonicotinic acid ≥95%
2-Fluoro-4-iodonicotinic acid

Hazard: Irritant/Light Sensitive

Formula: C₆H₃FINO₂
MDL Number: MFCD03095310
CAS Number: 884494-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR