2-Bromo-9,10-bis(2-naphthalenyl)anthracene ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-9,10-di(2-naphthyl)anthracene

073689-1G 073689-5G
102531-498EA 569.23 USD
102531-498 102531-500
2-Bromo-9,10-bis(2-naphthalenyl)anthracene ≥95%
2-Bromo-9,10-bis(2-naphthalenyl)anthracene

Hazard: Irritant

Formula: C₃₄H₂₁Br
MW: 509.4400 g/mol
MDL Number: MFCD11045064
CAS Number: 474688-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR