2,6-Dibromophenol ≥95%

Supplier: Matrix Scientific

073686-100G 073686-25G
102531-484EA 290.77 USD
102531-484 102663-954
2,6-Dibromophenol ≥95%
2,6-Dibromophenol

Hazard: Irritant

Formula: C₆H₄Br₂O
MDL Number: MFCD00002152
CAS Number: 608-33-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR