Methyl-4-formylbenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl terephthalaldehydate, Methyl 4-formylbenzoate

073685-100G 073685-500G
102531-476EA 69.23 USD
102531-476 102531-478
Methyl-4-formylbenzoate ≥95%
Methyl-4-formylbenzoate

Hazard: Irritant

Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 265°C
Melting Pt: 59-63°C
Flash Pt: 128°C
MDL Number: MFCD00006950
CAS Number: 1571-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR