trans-4-Aminoadamantan-1-ol hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
trans-1-Amino-4-hydroxyadamantane hydrochloride, trans-4-Amino-1-adamantanol hydrochloride
073684-1G
073684-5G
073684-10G
102531-468EA
533.85
USD
102531-468
102531-470
102531-472
trans-4-Aminoadamantan-1-ol hydrochloride ≥95%
trans-4-Aminoadamantan-1-ol hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₈ClNO MW: 203.7100 g/mol |
MDL Number:
MFCD11099880 CAS Number: 62075-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...