7-Bromo-6-chloroquinazolin-4(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 7-bromo-6-chloro-3,4-dihydroquinazolin-4-one

073683-1G 073683-5G 073683-25G
102531-462EA 15.38 USD
102531-462 102531-464 102833-650
7-Bromo-6-chloroquinazolin-4(3H)-one ≥95%
7-Bromo-6-chloroquinazolin-4(3H)-one

Hazard: Irritant

Formula: C₈H₄BrClN₂O
MW: 259.4900 g/mol
MDL Number: MFCD10000555
CAS Number: 17518-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR