3-Fluoro-o-anisidine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-2-methoxyaniline, 3-Fluoro-2-anisidine, 2-Amino-6-fluoroanisole

073682-1G 073682-5G 073682-10G
102531-456EA 143.08 USD
102531-456 102531-458 102531-460
3-Fluoro-o-anisidine ≥95%
3-Fluoro-o-anisidine

Hazard: Irritant

Formula: C₇H₈FNO
MDL Number: MFCD00007759
CAS Number: 437-83-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR