6-Aminopyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

073681-250MG 073681-1G 073681-5G
102531-450EA 90.77 USD
102531-450 102531-452 102531-454
6-Aminopyrazine-2-carboxylic acid ≥95%
6-Aminopyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
MDL Number: MFCD10697804
CAS Number: 61442-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR