3-Oxo-3,4-dihydropyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxypyrazine-2-carboxylic acid

073680-1G 073680-5G
102531-444EA 61.54 USD
102531-444 102531-446
3-Oxo-3,4-dihydropyrazine-2-carboxylic acid ≥95%
3-Oxo-3,4-dihydropyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C5H4N2O3
MW: 140.0969 g/mol
MDL Number: MFCD00235136
CAS Number: 20737-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR