5-Methyl-2-phenyl-1,2-dihydropyrazol-3-one ≥95%

Supplier: Matrix Scientific

073679-1G 073679-5G
102531-440EA 307.69 USD
102531-440 102531-442
5-Methyl-2-phenyl-1,2-dihydropyrazol-3-one ≥95%
5-Methyl-2-phenyl-1,2-dihydropyrazol-3-one

Hazard: Irritant

Formula: C10H10N2O
MDL Number: MFCD00127930
CAS Number: 19735-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR